Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

414999

Sigma-Aldrich

3,4-Epoxytetrahydrothiophene-1,1-dioxide

97%

Synonym(s):

3,4-Epoxysulfolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3S
CAS Number:
Molecular Weight:
134.15
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

145-150 °C (lit.)

SMILES string

O=S1(=O)CC2OC2C1

InChI

1S/C4H6O3S/c5-8(6)1-3-4(2-8)7-3/h3-4H,1-2H2

InChI key

SZAIAWVGWTXVMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to prepare sulfolene derivatives and acyclic polyenes used in natural product synthesis. For a synthesis, see Tetrahedron Lett.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 114-114 (1991)
Tetrahedron Letters, 2609-2609 (1971)
Journal of the Chemical Society. Perkin Transactions 1, 1335-1335 (1972)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service