Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

240230

Sigma-Aldrich

Phthalimide

≥99%

Synonym(s):

1,3-Dihydro-1,3-dioxoisoindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
Beilstein:
118522
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

mp

232-235 °C (lit.)

solubility

water: slightly soluble(lit.)

SMILES string

O=C1NC(=O)c2ccccc12

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)

InChI key

XKJCHHZQLQNZHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phthalimide was used in the synthesis of 6-amino agarose (AA) by Mitsunobu-inspired microwave mediated method.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alessio Pierini et al.
Journal of veterinary science, 21(5), e60-e60 (2020-10-06)
Tumor-associated neoangiogenesis is a crucial target for antitumor therapies. Thalidomide (TAL) is a promising anti-neoangiogenetic drug that has recently been used in the treatment of several malignancies in dogs. The aim of the study was to assess the pharmacokinetics of
Stalin Kondaveeti et al.
Carbohydrate polymers, 106, 365-373 (2014-04-12)
A facile 6-aminoagarose (AA) mediated synthesis of new fluorogenic amides of agarose with nicotinic (AA-NA) and picolinic acids (AA-PA) employing carbodiimide chemistry have been described. 6-Amino agarose (AA) was synthesized in a facile Mitsunobu-inspired microwave mediated method involving the reaction
Ive Hermans et al.
Physical chemistry chemical physics : PCCP, 10(8), 1125-1132 (2008-02-14)
Abstraction of hydrogen atoms by pthalimide-N-oxyl radicals is an important step in the N-hydroxyphthalimide catalyzed autoxidation of hydrocarbons. In this contribution, the temperature dependency of this reaction is evaluated by a detailed transition state theory based kinetic analysis for the
Jean L Santos et al.
Bioorganic & medicinal chemistry, 17(11), 3795-3799 (2009-05-12)
New phthalimide derivatives were easily prepared through condensation of phthalic anhydride and selected amines with variable yields (70-90%). All compounds (3a-l) were evaluated against Mycobacterium tuberculosis H(37)Rv using Alamar Blue susceptibility. The compounds 3c, 3i, and 3l have the minimum
Clarina I Manley-King et al.
Bioorganic & medicinal chemistry, 19(16), 4829-4840 (2011-07-23)
Literature reports that isatin as well as C5- and C6-substituted isatin analogues are reversible inhibitors of human monoamine oxidase (MAO) A and B. In general, C5- and C6-substitution of isatin leads to enhanced binding affinity to both MAO isozymes compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service