Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.07303

Sigma-Aldrich

Phthalimide

for synthesis

Synonym(s):

Phthalimide, Phthalic acid imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO2
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-603-3
NACRES:
NA.22

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

autoignition temp.

>500 °C

potency

>10000 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

pH

3.8 (0.6 g/L in H2O)

bp

310 °C/1013 hPa

mp

234-238 °C (sublimed)

transition temp

flash point 150 °C

solubility

0.6 g/L

density

1.21 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N1C(=O)c2c(cccc2)C1=O

InChI

1S/C8H5NO2/c10-7-5-3-1-2-4-6(5)8(11)9-7/h1-4H,(H,9,10,11)

InChI key

XKJCHHZQLQNZHY-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 233 °C
Melting range (upper value): ≤ 236 °C
Identity (IR): passes test

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service