Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.03600

Sigma-Aldrich

1,3-Dibromo-5,5-dimethylhydantoin

for synthesis

Synonym(s):

1,3-Dibromo-5,5-dimethylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6Br2N2O2
CAS Number:
Molecular Weight:
285.92
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-030-9
NACRES:
NA.22

Quality Level

form

powder

autoignition temp.

550 °C

potency

250 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

mp

186.0-192.0 °C

solubility

1 g/L

density

1.00 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

BrN1C(C(=O)N(C1=O)Br)(C)C

InChI

1S/C5H6Br2N2O2/c1-5(2)3(10)8(6)4(11)9(5)7/h1-2H3

InChI key

VRLDVERQJMEPIF-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service