Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

737135

Sigma-Aldrich

Dibromoisocyanuric acid

96%

Synonym(s):

1,3-Dibromo-1,3,5-triazine-2,4,6(1H,3H,5H)-trione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3HBr2N3O3
CAS Number:
Molecular Weight:
286.87
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

305-310 °C

storage temp.

2-8°C

SMILES string

BrN1C(=O)NC(=O)N(Br)C1=O

InChI

1S/C3HBr2N3O3/c4-7-1(9)6-2(10)8(5)3(7)11/h(H,6,9,10)

InChI key

HHBCEKAWSILOOP-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service