Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.03038

Sigma-Aldrich

Dicyclopentadiene

(stabilised) for synthesis

Synonym(s):

Dicyclopentadiene, Cyclopentadiene dimer, 4,7-Methano-3a,4,7,7a-tetrahydroindene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
MDL number:
UNSPSC Code:
12352002
EC Index Number:
201-052-9
NACRES:
NA.22

vapor pressure

3 hPa ( 20 °C)

Quality Level

Assay

≥90.0% (GC)

form

liquid

autoignition temp.

503 °C

potency

353 mg/kg LD50, oral (Rat)
4940 mg/kg LD50, skin (Rabbit)

expl. lim.

0.8-6.3 % (v/v)

kinematic viscosity

6 cSt(20 °C)

bp

166 °C/1013 hPa

mp

11-13 °C

density

0.98 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C10H12/c1-2-9-7-4-5-8(6-7)10(9)3-1/h1-2,4-5,7-10H,3,6H2

InChI key

HECLRDQVFMWTQS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 90.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.975 - 0.985
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

MOST offers controlled solar energy harvesting and storage, addressing global energy demands with improved storage techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service