Skip to Content
MilliporeSigma
All Photos(3)

Documents

C99205

Sigma-Aldrich

Cycloheptatriene

95%

Synonym(s):

Tropiliden, Tropilidene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8
CAS Number:
Molecular Weight:
92.14
Beilstein:
506066
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.519 (lit.)

bp

116-117 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1C=CC=CC=C1

InChI

1S/C7H8/c1-2-4-6-7-5-3-1/h1-6H,7H2

InChI key

CHVJITGCYZJHLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Contains 0.02%-0.15% BHT

Application

Cycloheptatriene can be used as a reactant to synthesize:      
  • 2-Hydroxyazulene via [2+2] cycloaddition reaction with dichloroketene, and subsequent ring expansion reaction with diazomethane.      
  • 7-Alkyl-bicyclo[4.2.1]nona-2,4,7-trienes by cobalt catalyzed [6+2] cycloaddition with terminal alkynes.     
  • (3,5-Cycloheptadienyl)diphenylphosphine ligand by reacting with diphenylphosphine.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Michael H Palmer et al.
The Journal of chemical physics, 153(5), 054301-054301 (2020-08-11)
The vacuum ultraviolet (VUV) spectrum for cyclohepta-1,3,5-triene up to 10.8 eV shows several broad bands, which are compared with electron impact spectra. Local curve fitting exposed groups of sharp vibrational peaks, which are assigned to Rydberg states. The vertical excitation
[2+ 2] Cycloaddition reaction of cycloheptatriene with dichloroketene. A novel and efficient synthetic strategy for the synthesis of 2-hydroxyazulene
Yokoyama R, et al.
Journal of the Chemical Society. Perkin Transactions 1, (18), 2257-2261 (2001)
Mathieu Achard et al.
Organic letters, 7(12), 2353-2356 (2005-06-04)
[reaction: see text] The CoI2(dppe)/Zn/ZnI2 system effectively catalyzes the [6 + 2] cycloaddition of cycloheptatriene with terminal alkynes to afford 7-alkyl-bicyclo[4.2.1]nona-2,4,7-trienes in fair to excellent yields. The catalyst proved to be tolerant toward functional groups such as ketone, sulfone, ester
(Cycloheptadienyl) diphenylphosphine: A Versatile Hybrid Ligand
Massard A, et al.
Organometallics, 31(3), 947-958 (2012)
Katharina Thum et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(7), 2513-2522 (2020-11-17)
The first intermolecular early main group metal-alkene complexes were isolated. This was enabled by using highly Lewis acidic Mg centers in the Lewis base-free cations (Me BDI)Mg+ and (tBu BDI)Mg+ with B(C6 F5 )4 - counterions (Me BDI=CH[C(CH3 )N(DIPP)]2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service