Skip to Content
MilliporeSigma
All Photos(1)

Documents

527408

Sigma-Aldrich

1,3-Hexadiene, mixture of cis and trans

95%

Synonym(s):

1-Ethyl-1,3-butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH=CH2
CAS Number:
Molecular Weight:
82.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.438 (lit.)

bp

72-76 °C (lit.)

density

0.714 g/mL at 25 °C (lit.)

SMILES string

CCC=CC=C

InChI

1S/C6H10/c1-3-5-6-4-2/h3,5-6H,1,4H2,2H3

InChI key

AHAREKHAZNPPMI-UHFFFAOYSA-N

Application


  • Ethylene-alt-alpha-Olefin Copolymers by Hydrogenation of Highly Stereoregular cis-1,4 Polydienes: A study on the synthesis and structural characterization of isotactic and syndiotactic polydienes, focusing on copolymers derived from cis-1,4 poly(1,3-hexadiene) (G Ricci et al., 2024).

  • Syndiotactic Alternating Ethylene/Propylene Copolymers Obtained Through Non-Catalytic Hydrogenation of Highly Stereoregular cis-1,4 Poly(1,3-diene)s: This article elaborates on the properties of ethylene/propylene copolymers derived from the hydrogenation of 1,3-diene polymers, including 1,3-hexadiene (G Ricci et al., 2017).

  • Novel cobalt dichloride complexes with hindered diphenylphosphine ligands: Research on catalytic systems for the polymerization of dienes including 1,3-hexadiene (G Ricci et al., 2019).

  • Synthesis and Reactivity of Pyridine (diimine) Molybdenum Olefin Complexes: A detailed look at ethylene dimerization and alkene dehydrogenation processes involving 1,3-hexadiene as an intermediate (MV Joannou et al., 2017).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laura Torrente-Murciano et al.
Frontiers in chemistry, 2, 37-37 (2014-07-02)
The first example of a tandem reaction involving double-bond migration in combination with telomerization is reported. Homogeneous and heterogeneous Ru catalysts were employed as isomerization catalysts, and telomerization was realized using a homogeneous Pd(0) precursor complex with a N-heterocyclic carbene
Natori et al.
Macromolecules, 31(15), 4687-4694 (1998-07-29)
The n-butyllithium (n-BuLi)/N,N,N',N'-tetramethylethylenediamine (TMEDA) system (with the molar ratio of TMEDA to n-BuLi higher than 4/4) has been found to polymerize 1,3-cyclohexadiene to produce "living" polymer having a narrow molecular weight distribution with well-controlled polymer chain length. The rate of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service