Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.02603

Sigma-Aldrich

Chloroacetone

(stabilised) for synthesis

Synonym(s):

Chloroacetone, 1-Chloro-2-propanone

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
92.52
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-161-1
NACRES:
NA.22

vapor pressure

42 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

610 °C

potency

100 mg/kg LD50, oral (Rat)
141 mg/kg LD50, skin (Rabbit)

expl. lim.

3.4 % (v/v)

pH

4.3 (20 °C, 124 g/L in H2O)

bp

119 °C/1013 hPa

mp

-44 °C

transition temp

flash point 36 °C

solubility

100 g/L

density

1.15 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C3H5ClO/c1-3(5)2-4/h2H2,1H3

InChI key

BULLHNJGPPOUOX-UHFFFAOYSA-N

General description

(stabilised) for synthesis

Application

Chloroacetone (stabilised) for synthesis. CAS No. 78-95-5, EC Number 201-161-1.

Analysis Note

Assay (GC, area%)≥ 94.5 % (a/a)Density (d 20 °C/ 4 °C)1.149 - 1.153Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service