Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.00236

Sigma-Aldrich

Cinnamoyl chloride

for synthesis

Synonym(s):

Cinnamoyl chloride, Cinnamic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOCl
CAS Number:
Molecular Weight:
166.60
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

form

crystals

mp

32-36 °C

transition temp

flash point >110 °C

density

1.16 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC\C=C\c1ccccc1

InChI

1S/C9H9Cl/c10-8-4-7-9-5-2-1-3-6-9/h1-7H,8H2/b7-4+

InChI key

IWTYTFSSTWXZFU-QPJJXVBHSA-N

Analysis Note

Assay (morpholine method): ≥ 98.0 %
Melting range (lower value): ≥ 32 °C
Melting range (upper value): ≤ 36 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service