Skip to Content
MilliporeSigma
All Photos(1)

Documents

71510-M

Millipore

D-Tube Dialyzer Maxi, MWCO 12-14 kDa

Synonym(s):

Large Dialyzer, MWCO 12-14 kDa Dialyzer, Maxi Dialyzer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

storage temp.

10-30°C

General description

The D-Tube Dialyzers are mainly used for dialysis and electroelution of proteins, RNA, DNA, and oligonucleotides from polyacrylamide or agarose gels. No syringes, microcentrifuges, or time-consuming steps are required to manipulate small sample volumes with the disposable, single-use tubes. Samples can be added and removed with the use of a standard laboratory pipet. This D-Tube dialyzer is designed with a volume capacity of 100-3000 μl. It contains 10 D-Tube dialyzers and one floating rack that can hold up to four D-Tube dialyzers. The membrane is ultra-clean and is free of sulfur and heavy metal.

Features and Benefits

  • Convenient dialyzers for buffer exchange and elimination of urea, detergents
  • Ethylenediamine tetra acetic acid (EDTA)-treated regenerated cellulose
  • Sample volume recovery >97%
  • Free of protease, RNAse, DNAse, and polymerase chain reaction (PCR) products

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service