Skip to Content
MilliporeSigma

Skip To

D6066

Dialysis sacks

Avg. flat width ~34 mm, MWCO 14,000 Da

Synonym(s):

Dialysis Bags, Dialysis Equipment

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pore size

14,000 Da MWCO

General description

Cellulose tubing; will retain proteins with M.W. > 12,000 Da
Inflated diameter approx. 21 mm (0.83 in.); pre-cut; open ended; dry unwashed; approx. length = 30 cm (12 in.)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D6191D9277D9402
description

Avg. flat width ~34 mm, MWCO 14,000 Da

description

Avg. flat width ~25 mm, MWCO 14,000 Da

description

avg. flat width 10 mm (0.4 in.)

description

avg. flat width 76 mm (3.0 in.)

pore size

14,000 Da MWCO

pore size

14,000 Da MWCO

pore size

-

pore size

-


Still not finding the right product?

Explore all of our products under Dialysis sacks




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library