Skip to Content
MilliporeSigma

71504-M

D-Tube Dialyzer Mini, MWCO 6-8 kDa

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

Novagen®

Quality Segment

storage temp.

10-30°C

Application

D-Tube Dialyzer Mini, MWCO 6-8 kDa has been used:
  • to dialyze pro matrix metalloprotease 1 (ProMMP-1) for MMP activation
  • in in vitro assay
  • for membrane hydration in electroelution

Features and Benefits

  • Compatible with most commercially available horizontal electrophoresis units
  • Efficient extraction (>90%) is achieved for 15 nt oligos and up to 80 kbp DNA fragments

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library