Skip to Content
MilliporeSigma

Skip To

6791

Vitamin B12

Synonym(s):

Vitamin B12, Cyanocobalamin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
$420.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25
eCl@ss:
34058004

$420.00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

RTECS - GG3750000
Merck USA index - 14, 10014

Quality Segment

assay

≥95% dry basis (UV)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

red-purple

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Co+3].[P](=O)([O-])(O[C@@H]6[C@H](O[C@@H]([C@@H]6O)[n]7c8c(nc7)cc(c(c8)C)C)CO)O[C@@H](CNC(=O)CC[C@@]1([C@H]([C@H]2N=C1C(=C3N=C(C([C@@H]3CCC(=O)N)(C)C)C=C4N=C([C@]([C@@H]4CCC(=O)N)(CC(=O)N)C)C(=C5[N-][C@@]2([C@]([C@@H]5CCC(=O)N)(CC(=O)N)C)C)C)C)CC(=O)N)C)

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;-1;+3/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1

InChI key

FDJOLVPMNUYSCM-WZHZPDAFSA-L

General description

Part of coenzyme needed for the synthesis of proteins, and nucleic acids and for the metabolism of carbohydrates. Plays a role in the synthesis of myelin. Used as an antipernicious anemia factor.

Preparation Note

Unstable in solution; reconstitute just prior to use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V66291.24592C3000000
assay

≥95% dry basis (UV)

assay

≥98%

assay

-

assay

-

form

powder

form

powder

form

solid

form

solid

solubility

water: 10 mg/mL

solubility

water: soluble 50 mg/mL, dark red

solubility

12 g/L

solubility

-

storage temp.

15-25°C

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

2-8°C

color

red-purple

color

red to brown

color

-

color

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

-


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters