Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616407

Sigma-Aldrich

TACE Substrate IV, Fluorogenic

Synonym(s):

TACE Substrate IV, Fluorogenic, TNF-α Convertase Enzyme Substrate IV, Fluorogenic, Abz-LAQAVRSSSR-Dpa, ADAM-17 Substrate IV, Fluorogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H93N23O20
Molecular Weight:
1444.51
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

yellow

solubility

DMSO: 5 mg/mL

fluorescence

λex ~320 nm
λem ~420 nm

shipped in

ambient

storage temp.

2-8°C

General description

An internally quenched, fluorogenic, 10-amino acid peptide substrate that is useful for monitoring TNF-α Convertase Enzyme (TACE) activity by a continuous fluorometric assay. Also useful for screening TACE inhibitors (kcat = 21.6 s-1 when the substrate concentration is half the Vmax, S0.5 = 342 µM). The substrate is cleaved at the Ala-Val amide bond. The substrate is not substantially cleaved by MMP-1, MMP-9, or MMP-13. Displays improved solubility in aqueous buffers. Excitation max: ~320 nm; Emission max: ~420 nm.
An internally-quenched fluorogenic decapeptide substrate that is useful for continuous monitoring of TACE activity and high-throughput screening of TACE inhibitors (kcat = 21.6 s-1 and substrate concentration at half the Vmax, S0.5 = 342 µM). The substrate is cleaved at Ala-Val amide bond. Acts as a poor substrate for MMP-1, MMP-9, and MMP-13. Displays better solubility in aqueous buffers.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fluorogenic decapeptide substrate that is useful for continuous monitoring of TACE activity
Product does not compete with ATP.
Reversible: no
kcat = 21.6 s-1 and substrate concentration at half the Vmax, S0.5 = 342 µM

Warning

Toxicity: Standard Handling (A)

Sequence

Abz-Leu-Ala-Gln-Ala-Val-Arg-Ser-Ser-Ser-Arg-Dpa

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Jin, G., et al. 2002. Anal. Biochem.302, 269.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service