Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

5.74846

Millipore

Buffered Peptone Water

ReadyStream® (Media bag), GranuCult® prime, according to ISO, FDA-BAM, EP, granular, pack of 3 × 10 L, pack of 10 × 3 L, for use with ReadyStream® System

Synonym(s):

Buffered, Buffered Peptone Solution, Concentrated media, GranuCult® Buffered peptone water media, ISO compliant BPW media, Peptone Water, Peptone Water (buffered), Phosphate-buffered, ReadyStream® Buffered Peptone Water, Salmonella testing media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.74

Agency

EP
FDA-BAM
ISO 11133
ISO 19250
ISO 21528
ISO 22964
ISO 6579
ISO 6887
according to ISO

Quality Level

form

granular

packaging

pack of 3 × 10 L
pack of 10 × 3 L

manufacturer/tradename

GranuCult® prime
ReadyStream® (Media bag)

storage condition

(Store dry and tightly closed)

technique(s)

microbiological culture: suitable

pH

7.0± 0.2 (25 °C)

application(s)

food and beverages

compatibility

for use with ReadyStream® System

storage temp.

15-25°C

suitability

nonselective for bacteria

Looking for similar products? Visit Product Comparison Guide

General description

The innovative ReadyStream® workflow allows convenient production of up to 100 L of pre-heated, ISO 11133 compliant BPW media in less than 20 minutes. ReadyStream® Buffered Peptone Water which contains our Granucult® media is shipped as a dry media bag that has been γ-irritated. Installed at the ReadyStream® system (RDYSTRM01), the media is first reconstituted with sterile filtered water to create a 10x concentration of the BPW media. Subsequently, the media is dispensed and diluted by the ReadyStream® system to obtain the 1x or 2x concentrated media. Thanks to this the 10-fold volume of the media stock, i.e., depending on media bag size 30 L and 100 L, can be easily prepared. Once reconstituted the bag can be used for 5 days.

Application

ReadyStream® Buffered Peptone Water is used in the testing of food and water samples. This product is suitable for the preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter, from food, animal feed, water, and other materials. The product is also suitable as a diluent for general use.

Other Notes

To access the ReadyStream® Data Matrix Generator Tool follow this link https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

Salmonella pass through the entire food chain from animal feed, primary production, and into households or food-service establishments.

See All

Related Content

The Revolution in Media Preparation - New solution for media preparation and dispensing for food testing labs

The Revolution in Media Preparation - New solution for media preparation and dispensing for food testing labs

The Revolution in Media Preparation - New solution for media preparation and dispensing for food testing labs

The Revolution in Media Preparation - New solution for media preparation and dispensing for food testing labs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service