Skip to Content
MilliporeSigma

RDYSTRM10

ReadyStream® Media Unit

For convenient preparation of large amount of food testing media in short time

Synonym(s):

Automatic Media Preparation Unit, Concentrated Media Preparator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
13102014
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

200 V / 240 V, 50-60 Hz

Quality Segment

manufacturer/tradename

ReadyStream®

system H × D × L

20.5 cm × 50.1 cm × 35.3 cm

application(s)

food and beverages

compatibility

for use with ReadyStream® System

General description

ReadyStream® Media Unit is an integral part of ReadyStream® system designed for instant and sterile media preparation.
It is a compact media preparation unit comprising:
  • Outlet connector for filter set (water sterile filtration)
  • Peristaltic pump (automatic dosing of concentrated media)
  • 4-way stopcock actuator (for combining the water from the filter set, and concentrated media from the media bag)

ReadyStream® Media Unit automatically reconstitutes the dehydrated granulated media (supplied separately with tubing set connection) with sterile filtered water (done by the instrument).
The prepared media can be used directly without autoclaving. The media is dispensed through the ReadyStream® Dispensing Unit with chosen media volume and temperature.
ReadyStream® System can prepare and dispense sterile media with minimal user intervention. The benchtop design saves laboratory space and time involved in critical experiments and daily routine.

Application

ReadyStream® Media Unit is used to prepare and dispense instant sterile media. The prepared media is ready to use for routine microbiological applications.

Features and Benefits

  • Ready-to-use media.
  • No Autoclaving.
  • 15-20 minutes of preparation time.
  • Benchtop system to save laboratory space vs. traditional media preparation process.
  • Large Media volume available on demand e.g. 100L from 10L concentrated stock media bag.

Reduced workflow, time, energy, and water consumption.

Other Notes

Components required but not included:
This catalog number doesnt include all components that are required for the installation of the entire ReadyStream® System. The following parts need to be ordered separately along with this product:
  • RDYSTRM20.
  • RDYSTRM30.
  • RDYSTRM40.
  • RDYSTRM90.
  • RDYSTRAIN01.
  • A country specific Power Cord adapted to local electrical requirements.
The indicated Price currently corresponds to the above catalog number only.
To access the ReadyStream® Data Matrix Generator Tool follow this link:https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library