Skip to Content
MilliporeSigma

RDYSTRM20

ReadyStream® Media Dispensing Unit

For convenient dispensing of large amount of food testing media in short time

Synonym(s):

Automatic Media Dispensing Unit, Concentrated Media Preparator

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
13102014
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

24 V / 24 V DC

Quality Segment

manufacturer/tradename

ReadyStream®

system H × D × L

46.6 cm × 35.3 cm × 23.1 cm

application(s)

food and beverages

compatibility

for use with ReadyStream® system

General description

ReadyStream® Media Dispensing Unit is designed to dispense sterile media prepared by the ReadyStream® Media Unit as part of the ReadyStream® System
The dispensing unit comprises the following parts:
  • Graduated Unit Stand (to adjust dispensing height).
  • Marked Positions for standard and large ReadyStream® bag holder.
  • Bag holder magnet.
  • Control Panel.

ReadyStream® Media Dispensing Unit dispenses small and pooled sample media volumes directly into sample bags or containers using pre-programmed or customer generated parameters. The prepared media can be used directly without autoclaving.
The ReadyStream® Media Dispensing Unit can also be used for dispensing sterile water based on the user need and requirement.
ReadyStream® System can prepare and dispense sterile media with minimal user intervention. The benchtop design saves laboratory space and time involved in critical experiments and daily routine.

Application

ReadyStream® Media Dispensing Unit is used to dispense sterile media into standard and large bags based on the application.

Features and Benefits

  • Height Adjuster screw.
  • Aseptic Dispensing.
  • Custom Programming available.
  • Only water dispensing feature.

Easy Maintenance.

Other Notes

Components required but not included: ·

This catalog number doesn’t include all components that are required for the installation of the entire ReadyStream® System.

The following parts need to be ordered separately along with this product:

  • RDYSTRM10.
  • RDYSTRM30.
  • RDYSTRM40.
  • RDYSTRM90.
  • RDYSTRAIN01. ·


A country specific Power Cord adapted to local electrical requirements. ·

The indicated Price currently corresponds to the above catalog number only.
To access the ReadyStream® Data Matrix Generator Tool follow this link https://www.sigmaaldrich.com/US/en/support/calculators-and-apps/readystream-data-matrix-generator

Legal Information

READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library