Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565852

Sigma-Aldrich

PTP Inhibitor XXXI, II-B08

The PTP Inhibitor XXXI, II-B08 controls the biological activity of PTP. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PTP Inhibitor XXXI, II-B08, CD45 Inhibitor V, FAP1 Inhibitor, LMWPTP Inhibitor, Lyp Inhibitor III, PTP1B Inhibitor IX, SHP1 Inhibitor IX, SHP2 Inhibitor VI, 3-(1-(3-(Biphenyl-4-ylamino)-3-oxopropyl)-1H-1,2,3-triazol-4-yl)-6-hydroxy-1-methyl-2-phenyl-1H-indole-5-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H27N5O4
Molecular Weight:
557.60
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

potency

5.2 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable indolo-salicylic acid compound that is shown to target SHP2 catalytic domain and inhibit SHP2-catalyzed pNPP hydrolysis (IC50 = 5.5 µM) in a reversible and non-competitive manner (Ki = 5.2 µM), exhibiting 2.6- to 5.7-fold selectivity over PTP1B, SHP1, FAP1, Lyp, CD45, and LMWPTP (IC50 = 14.3, 15.7, 20.3, 25.0, 30.0, and 31.1 µM, respectively) and >9-fold selectivity over Cdc14A, HePTP, LAR, PTPα, and VHR (IC50 >50 µM). Effectively blocks hyperproliferative activity of human hematopoietic progenitor cells (Effective conc. = 10 µM) due to SHP2 mutations (D61Y and E76K) or GM-CSF stimulation, as well as c-kit-dependent (via ligand stimulation or D814V mutation), SHP2-mediated myeloid proliferation (Effective conc. = 5 to 75 µM). Dual PI-3K and SHP2 inhibition by LY294002 (Cat. Nos. 440202 & 440204) and II-B08 (10 mg/kg and 50 mg/kg, respectively; daily i.p.) is shown to greatly prolong the survival rate in a D814V KIT-expressing 32D-based murine MPD (myeloproliferative disease) model (0% survival rate on day 23, 27, 28, respectively, with DMSO, LY294002, II-B08 treatment alone; 20% survival rate on day 65 with combined treatment) in vivo.
A cell-permeable indolo-salicylic acid compound that is shown to target SHP2 catalytic domain and inhibit SHP2-catalyzed pNPP hydrolysis (IC50 = 5.5 µM) in a reversible and non-competitive manner (Ki = 5.2 µM), exhibiting 2.6- to 5.7-fold selectivity over PTP1B, SHP1, FAP1, Lyp, CD45, and LMWPTP (IC50 = 14.3, 15.7, 20.3, 25.0, 30.0, and 31.1 µM, respectively). Effectively blocks SHP2-mediated human hematopoietic progenitor cell hyperproliferation (10 µM) due to SHP2 mutations (D61Y and E76K) or GM-CSF stimulation, as well as c-kit-dependent (due to D814V mutation or via ligand stimulation), SHP2-mediated myeloid proliferation (5 to 75 µM). Dual PI-3K and SHP2 inhibition by LY294002 (Cat. Nos. 440202 & 440204) and II-B08 (10 mg/kg and 50 mg/kg, respectively; daily i.p.) is shown to greatly prolong the survival rate of mice bearing D814V KIT-expressing 32D cells in vivo.

Biochem/physiol Actions

Primary Target
SHP2
Reversible: yes
Secondary Target
Other PTPs
Target IC50: 5.5, 9.0 and 10.8 &micro

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Mali, R.S., et al. 2012. Blood120, 2669.
Zhang, X., et al. 2010. J. Med. Chem.53, 2482.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service