Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1465

Sigma-Aldrich

UNC0379 trifluoroacetate salt

≥98% (HPLC)

Synonym(s):

6,7-Dimethoxy-2-(pyrrolidin-1-yl)-N-(5-(pyrrolidin-1-yl)-pentyl)quinazolin-4-amine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H35N5O2 · xC2HF3O2
CAS Number:
Molecular Weight:
413.56 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

COC1=C(OC)C=C(N=C(N2CCCC2)N=C3NCCCCCN4CCCC4)C3=C1

InChI

1S/C23H35N5O2/c1-29-20-16-18-19(17-21(20)30-2)25-23(28-14-8-9-15-28)26-22(18)24-10-4-3-5-11-27-12-6-7-13-27/h16-17H,3-15H2,1-2H3,(H,24,25,26)

InChI key

WEXCGGWTIDNVNT-UHFFFAOYSA-N

Biochem/physiol Actions

SETD8 expression is observed in cancers such as bladder cancer, chronic myelogenous leukemia, hepatocellular carcinoma, non‐small‐cell lung carcinoma, prostate cancer, small‐cell lung carcinoma. Therefore inhibition of SETD8 (SET‐domain containing protein) by UNC0379 might prevent the progression of cancer.
UNC0379 is the first substrate-competitive inhibitor of the lysine methyltransferase SETD8, the only known methyltransferase that catalyzes monomethylation of histone H4 lysine 20 (H4K20). HK420 methylation has been implicated in the regulation of a variety of biological processes including the DNA damage response. UNC0379 is selective for SETD8 over 15 other methyltransferases, with an IC50 value of 7.3 μM for SETD8 compared to IC50 vlaues over 100 μM for other methyltransferases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivia G Dobbs et al.
BMC biology, 21(1), 175-175 (2023-08-15)
Cip1-interacting zinc finger protein 1 (CIZ1) forms RNA-dependent protein assemblies that stabilise epigenetic state, notable at the inactive X chromosome in females. CIZ1 has been linked with a range of human cancers and in mice genetic deletion of CIZ1 manifests
Dysregulation of protein methyltransferases in human cancer: an emerging target class for anticancer therapy.
Hamamoto R and Nakamura Y
Cancer Science, 107(4), 377-384 (2016)
Miku Wada et al.
Biomolecules, 10(12) (2020-12-20)
The histone methyltransferase SETD8, which methylates the lysine 20 of histone H4 (H4K20), is reportedly involved in human carcinogenesis along with nonhistone proteins such as p53. However, its expression profiles and functions in the context of high-grade serous ovarian carcinoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service