Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

565770

Sigma-Aldrich

DAPT

≥98% (HPLC), lyophilized, γ-secretase inhibitor, Calbiochem®

Synonym(s):

γ-Secretase Inhibitor IX, DAPT, N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26F2N2O4
CAS Number:
Molecular Weight:
432.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Product Name

γ-Secretase Inhibitor IX, Gamma-Secretase Inhibitor IX - CAS 208255-80-5, is a cell-permeable inhibitor of γ-secretase (Aβtotal IC₅₀ = 115 nM, Aβ42 IC₅₀ = 200 nM).

Quality Level

Assay

≥98% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1cc(cc(c1)CC(=O)N[C@@H](C)C(=O)N[C@@H](c2ccccc2)C(=O)OC(C)(C)C)F

InChI

1S/C23H26F2N2O4/c1-14(26-19(28)12-15-10-17(24)13-18(25)11-15)21(29)27-20(16-8-6-5-7-9-16)22(30)31-23(2,3)4/h5-11,13-14,20H,12H2,1-4H3,(H,26,28)(H,27,29)/t14-,20-/m0/s1

InChI key

DWJXYEABWRJFSP-XOBRGWDASA-N

General description

A cell-permeable dipeptide that inhibits γ-secretase activity and suppresses Aβ production (Aβ total IC50 = 115 nM; Aβ42 IC50 = 200 nM). Reported to be functionally active in both HEK293 cells overexpressing human APP751 and neuronal cultures, however, it does not affect the secretion of amyloid-b precursor protein (APP). Effective in acutely lowering Ab in APPV717F-transgenic mice. Also effective in inhibiting the production of an ~6 kDa C-terminal cleavage product (CTFg) in HEK293 cells stably transfected with the Swedish mutant, βAPP695. Reported to reduce extracellular Aβ plaques and intracellular Aβ accumulation in 3xTgAD transgenic mice.
A cell-permeable dipeptide that suppresses Aβ production by blocking γ-secretase (Aβtotal IC50 = 115 nM, Aβ42 IC50 = 200 nM). Prevents Aβ34 generation in murine N2a cells and hAPP/hBACE-1 transfected human HEK 293 cells. Reported to be functionally active in both HEK 293 cells and neuronal cultures without affecting the secretion of amyloid-β precursor protein (APP). Also shown to lower Aβ levels acutely in APP-transgenic mice. Reported to reduce extracellular Aβ plaques and intracellular Aβ accumulation in 3xTgAD transgenic mice. A 25 mM (5 mg/462 µl) solution of γ-Secretase Inhibitor IX (Cat. No. 565784) in DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 115 nM suppressing Aβtotal production; 200 nM suppressing Aβ42 production

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Morohashi, Y., et al. 2006. J. Bio. Chem.Vol. 281, 14670.
Nishitomo, K. et al. 2006 J. Neurochem.99, 1555.
Oddo, S., et al. 2004. Neuron43, 321.
Dovey, H.F., et al. 2001. J. Neurochem.76, 173.
Sastre, M., et al. 2001. EMBO Rep.2, 835.
Vandermeeren, M., et al. 2001. Neurosci. Lett.315, 145.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Bing et al.
Developmental neurobiology, 83(5-6), 157-166 (2023-07-11)
There is obvious sexual dimorphism in the song control system of songbirds. In the higher vocal center (HVC), cell proliferation and neuronal differentiation contribute to the net addition of neurons. However, the mechanism underlying these changes is unclear. Given that
Shuyang Cai et al.
Nature communications, 15(1), 226-226 (2024-01-04)
Hematopoietic stem and progenitor cells generate all the lineages of blood cells throughout the lifespan of vertebrates. The emergence of hematopoietic stem and progenitor cells is finely tuned by a variety of signaling pathways. Previous studies have revealed the roles
Christiane Volbracht et al.
Analytical biochemistry, 387(2), 208-220 (2009-05-21)
Amyloid-beta peptide (Abeta), a putatively causative agent of Alzheimer's disease (AD), is proteolytically derived from beta-amyloid precursor protein (APP). Here we describe cellular assays to detect the activity of the key protease beta-site of APP cleaving enzyme 1 (BACE1) based
Maike Marczenke et al.
Development (Cambridge, England), 148(21) (2021-10-27)
Growth arrest-specific 1 (GAS1) acts as a co-receptor to patched 1, promoting sonic hedgehog (SHH) signaling in the developing nervous system. GAS1 mutations in humans and animal models result in forebrain and craniofacial malformations, defects ascribed to a function for
Emanuele Azzoni et al.
Cell reports, 37(11), 110103-110103 (2021-12-16)
Hematopoietic stem cells (HSCs) emerge during development from the vascular wall of the main embryonic arteries. The onset of circulation triggers several processes that provide critical external factors for HSC generation. Nevertheless, it is not fully understood how and when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service