Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

361566

Sigma-Aldrich

GSK-3 Inhibitor IV, SB-216763

Synonym(s):

GSK-3 Inhibitor IV, SB-216763, SB-216763

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12Cl2N2O2
CAS Number:
Molecular Weight:
371.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

orange

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C19H12Cl2N2O2/c1-23-9-13(11-4-2-3-5-15(11)23)17-16(18(24)22-19(17)25)12-7-6-10(20)8-14(12)21/h2-9H,1H3,(H,22,24,25)

InChI key

JCSGFHVFHSKIJH-UHFFFAOYSA-N

General description

A cell-permeable (aryl,indole) maleimide compound that acts as a potent, selective, ATP-competitive inhibitor of GSK-3 activity (Ki = 9.1 nM for GSK-3α). At 10 µM concentration, does not significantly inhibit a panel of 24 other protein kinases tested. Reported to stimulate glycogen synthesis in Chang human liver cells (EC50 = 3.6 µM) and induce transcription of β-catenin-LEF/TCF (lymphoid enhancer factor/T cell factor) regulated reporter gene in HEK293 human embryonic kidney cell lines. Offers protection against polyglutamine-induced death in SK-N-SH and COS-7 cells and attenuates hypoxia-induced apoptosis in VSMC and COS-7 cells. Shown to reduce phosphoenolpyruvate carboxykinase (PEPCK) and glucose-6-phosphatase (G6Pase) expression in hepatoma cells. Promotes the survival of CLL lymphocytes via Wnt/β-catenin-mediated transcription activation. Exhibits delayed cardioprotection in vivo in a rat model via a KATP (ATP-sensitive potassium channels) and MPTP (mitochondrial permeability transition pore)-dependent mechanism at reperfusion.
A cell-permeable (aryl,indole) maleimide compound that acts as a potent, selective, ATP-competitive inhibitor of GSK-3 activity (Ki = 9.1 nM for GSK-3α). At 10 µM concentration, does not significantly inhibit a panel of 24 other protein kinases tested. Reported to stimulate glycogen synthesis in Chang human liver cells (EC50 = 3.6 µM) and induce transcription of β-catenin-LEF/TCF (lymphoid enhancer factor/T cell factor) regulated reporter gene in HEK293 human embryonic kidney cell lines. Offers protection against polyglutamine-induced death in SK-N-SH and COS-7 cells and attenuates hypoxia-induced apoptosis in VSMC and COS-7 cells. Shown to reduce phosphoenolpyruvate carboxykinase (PEPCK) and glucose-6-phosphatase (G6Pase) expression in hepatoma cells. Promotes the survival of CLL lymphocytes via Wnt/β-catenin-mediated transcription activation. Exhibits delayed cardioprotection in vivo in a rat model via a KATP (ATP-sensitive potassium channels) and MPTP (mitochondrial permeability transition pore)-dependent mechanism at reperfusion.

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Gross, E., et al. 2008. Am. J. Phy. Heart Circ Physiol.294, H1497.
Bain, J., et al. 2007. Biochem. J.408, 297.
Lu, D., et al. 2004. PNAS.101, 3118.
Carmichael, J., et al. 2002. J. Biol. Chem.277, 33791.
Culbert, A.A., et al. 2001. FEBS Lett.507, 288.
Lochhead, P.A., et al. 2001. Diabetes50, 937.
Cross, D.A., et al. 2001. J. Neurochem.77, 94.
Coghlan, M.P., et al. 2000. Chem. Biol.7, 793.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service