Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C20H30N2O4
Molecular Weight:
362.46
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥95% (HPLC)
Form:
solid
Storage condition:
OK to freeze, protect from light
Pricing and availability is not currently available.
Quality Segment
assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, protect from light
color
white
solubility
DMSO: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
General description
A cell-permeable, reversible dipeptide aldehyde that reduces Aβ production by blocking γ-secretase activity in vitro (Aβ40 IC50 = 7.9 µM; Aβ42 IC50 = 7.6 µM) and in cultured CHO 2b-7 cells that stably overexpress APP695 (Aβ40 IC50 = 11.5 µM; Aβ42 IC50 = 8.3 µM). Also blocks the generation of CTFγ (γ-secretase-generated carboxyl-terminal fragment). Does not affect the formation of amyloid-β-precursor protein (APP).
Biochem/physiol Actions
Cell permeable: yes
Primary Target
Aβ40 in vitro
Aβ40 in vitro
Product does not compete with ATP.
Reversible: yes
Target IC50: 7.9 µM for Aβ40; 7.6 µM for Aβ42 in vitro
Packaging
Packaged under inert gas
Other Notes
Weijzen, S., et al. 2002. Nat. Med.8, 979.
McLendon, C., et al. 2000. FASEB J.14, 2383.
McLendon, C., et al. 2000. FASEB J.14, 2383.
Z-Ile-Leu-CHO
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form lyophilized | form solid | form solid |
| assay ≥95% (HPLC) | assay ≥98% (HPLC) | assay ≥95% (HPLC) | assay ≥95% (HPLC) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® | manufacturer/tradename Calbiochem® |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| storage condition OK to freeze, protect from light | storage condition OK to freeze, protect from light | storage condition OK to freeze | storage condition OK to freeze, protect from light |
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



