Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5.34329

Sigma-Aldrich

H2S Fluorescent Probe, P3

Synonym(s):

H2S Fluorescent Probe, P3, ( E)-2-(3-(6-(2-Hydroxyethylamino)naphthalen-2-yl)-3-oxoprop-1-enyl)-3,5-dimethoxybenzaldehyde, Hydrogen Sulfide Fluorescent Probe, P3, Probe P3, (E)-2-(3-(6-(2-Hydroxyethylamino)naphthalen-2-yl)-3-oxoprop-1-enyl)-3,5-dimethoxybenzaldehyde, Hydrogen Sulfide Fluorescent Probe, P3, Probe P3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23NO5
Molecular Weight:
405.44
UNSPSC Code:
12352202
NACRES:
NA.28

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 100 mg/mL

fluorescence

λex 375 nm
λem 505 nm

storage temp.

2-8°C

General description

A cell-permeable, non-toxic, non-fluorescent naphthalenyl-dimethoxybenzaldehyde enone compound that acts as a highly specific, sensitive, fast-responsive H2S fluoroprobe (signal saturation at ≤ 5 min with a detection limit of 50 nM). Dose-dependently detects hydrogen sulfide levels under physiological conditions and readily fluoresces. Displays little to no reactivity towards GSH, Cys, H2O2, citrate and iodide (Hepes pH 7.4, containing 10% FBS). Also shown to be useful as a two-photon probe for imaging endogeneous H2S levels in HeLa cells. A thiol specific fluorescent probe is also available (Cat. No. 595504).Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
hydrogen sulfide
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Singha, S., et al. 2015. Anal. Chem.87, 1188.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service