Skip to Content
MilliporeSigma

SML3326

WSP5

≥90% (HPLC), fluorescent hydrogen sulfide probe, powder

Synonym(s):

1,1′-(3-Oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-3′,6′-diyl) bis[2-(2-pyridinyldithio)benzoate], Benzoic acid, 2-(2-pyridinyldithio)-, 1,1′-(3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-3′,6′-diyl) ester, H2S fluorescent probe WSP5, Hydrogen sulfide fluorescent probe WSP-5, WSP-5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C44H26N2O7S4
CAS Number:
Molecular Weight:
822.95
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥90% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

WSP5, ≥90% (HPLC)

Quality Segment

assay

≥90% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=C(SSC2=NC=CC=C2)C=CC=C1)OC3=CC=C4C(OC5=CC(OC(C6=C(SSC7=NC=CC=C7)C=CC=C6)=O)=CC=C5C48OC(C9=C8C=CC=C9)=O)=C3

InChI

1S/C44H26N2O7S4/c47-41(30-12-2-5-15-37(30)54-56-39-17-7-9-23-45-39)50-27-19-21-33-35(25-27)52-36-26-28(20-22-34(36)44(33)32-14-4-1-11-29(32)43(49)53-44)51-42(48)31-13-3-6-16-38(31)55-57-40-18-8-10-24-46-40/h1-26H

InChI key

DSMMZTASSIAQPQ-UHFFFAOYSA-N

Biochem/physiol Actions

Cell-permeable, highly selective, turn-on fluorescent probe for hydrogen sulfide (H2S)
WSP5 is a cell-permeable fluoresceinyl pyridine disulfide compound that acts as a highly selective, fast- and time-dependent nucleophilic substitution–cyclization-based H2S sensitive fluorescent turn-on live cells probe (60-fold; ƛex = 502 nm & ƛem = 525 nm; ɸ = 0.20 & a detection limit = 47 nM). WSP5 is reported to function over biological pH ranges and insensitive towards bio-thiols, various reactive sulfur species and esterases.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
SML3326-25MG04065269131632
SML3326-5MG04065269131649