Skip to Content
MilliporeSigma

SCT211

BioTracker Green Sulfane Sulfur live cell dye

Live Cell imaging probe for sulfane sulfur species including persulfide (R-S-SH), polysulfide (R-S-S<sub>n</sub>-S-R) and ploysulfide (H<sub>2</sub>S<sub>n</sub>).

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

lyophilized

application(s)

cell analysis

detection method

fluorometric

General description

Intracellularly abundant sulfane sulfur species including persulfide (R-S-SH), polysulfide (R-S-Sn-S-R) and polysulfide (H2Sn) play important roles to maintain intracellular reducing environments. BioTracker Green Sulfane Sulfur Live Cell Dye is a FRET-based fluorescence probe to specifically detect the intracellular sulfane sulfur species. It shows only weak fluorescence without sulfane sulfur, and reversibly fluoresces in response to micromolar concentrations of sulfane sulfur. Its reaction is highly specific and has minimal to no reactivity with H2S, cysteine residues, and sulfur oxides. The dye is a cell permeable diacetylated form which is hydrolyzed by intracellular esterases to generate a metabolite which is retained within the cells. Thus it is suitable to monitor intracellular concentration of sulfane sulfur via live-cell imaging.Spectral Properties:Absorbance (max): 495nmEmission (max): 525nmResearch Category:Live Cell Dye

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library