Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

516726

Millipore

Phenol/Chloroform/Isoamyl Alcohol, 25:24:1 (v/v), Molecular Biology Grade

A 25:24:1 (v/v) mixture of phenol, chloroform and isoamyl alcohol, pH 6.7. Suitable for DNA and RNA applications.

Synonym(s):

Organic Solvent Mix for DNA/RNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.21
grade:
Molecular Biology

grade

Molecular Biology

Quality Level

Assay

≥99% phenol basis (titration)
>99% chloroform basis (HPLC)

form

liquid (Two-layer)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless

shipped in

ambient

storage temp.

2-8°C

General description

A 25:24:1 (v/v) mixture of phenol, chloroform and isoamyl alcohol, pH 6.7. Suitable for DNA and RNA applications. Prior to equilibration with the Tris-based equilibration buffer (pH ~6.7), the mix is ideal for extraction of proteins and lipids for RNA purification. Following equilibration with the Tris-based equilibration buffer (pH ~8.0), is ideal for extraction of proteins and lipids for DNA purification. Absorbance: ≤0.2 at 300 nm; and ≤0.1 at 405 nm and 510 nm. Volume 400 ml. Supplied with 26 ml Tris-based equilibration buffer.

Warning

Toxicity: Toxic (F)

Reconstitution

Following equilibration, refrigerate (4°C).

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyan You et al.
PloS one, 14(9), e0222292-e0222292 (2019-09-26)
The giant panda (Ailuropoda melanoleuca) is a native species to China. They are rare and endangered and are regarded as the 'national treasure' and 'living fossil' in China. For the time being, there are only about 2500 giant pandas in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service