Skip to Content
MilliporeSigma

Skip To

77618

Phenol – chloroform – isoamyl alcohol mixture

BioUltra, Molecular Biology, 49.5:49.5:1

Phenol – chloroform – isoamyl alcohol mixture
1 of 1 reviewers took part in a promotion

Synonym(s):

phenol:Chloroform:Isoamyl Alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
NACRES:
NA.07
PubChem Substance ID:
UNSPSC Code:
12352401
MDL number:
Grade:
Molecular Biology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

product line

BioUltra

form

liquid

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

pH

7.7-8.3(H2O-phase, after extraction with H2O, 1:1)

density

1.29 g/mL at 20 °C

absorption

cut-off at 360 nm

storage temp.

2-8°C

SMILES string

ClC(Cl)Cl.CC(C)CCO.Oc1ccccc1

InChI

1S/C6H6O.C5H12O.CHCl3/c7-6-4-2-1-3-5-6;1-5(2)3-4-6;2-1(3)4/h1-5,7H;5-6H,3-4H2,1-2H3;1H

InChI key

ZYWFEOZQIUMEGL-UHFFFAOYSA-N

Application


  • Simultaneous quantification of multiple RNA cargos co-loaded into nanoparticle-based delivery systems.: This study highlights the use of a phenol-chloroform-isoamyl alcohol mixture for the extraction and quantification of RNA cargos from nanoparticles, facilitating accurate delivery system analysis (Lokras et al., 2022).

  • Analysis of Circulating microRNAs and Their Post-Transcriptional Modifications in Cancer Serum by On-Line Solid-Phase Extraction-Capillary Electrophoresis-Mass Spectrometry.: This research utilized the phenol-chloroform-isoamyl alcohol mixture to extract microRNAs from serum samples, providing insights into cancer biomarkers (Pero-Gascon et al., 2018).

  • The application of alkaline lysis and pressure cycling technology in the differential extraction of DNA from sperm and epithelial cells recovered from cotton swabs.: The study demonstrates the effectiveness of using phenol-chloroform-isoamyl alcohol for DNA extraction in forensic applications, improving the analysis of mixed samples (Nori and McCord, 2015).

  • The critical role of DNA extraction for detection of mycobacteria in tissues.: This article emphasizes the necessity of phenol-chloroform-isoamyl alcohol extraction in accurately detecting mycobacterial DNA in tissue samples, enhancing diagnostic precision (Radomski et al., 2013).

Physical form

mixture of phenol; chloroform; isoamyl alcohol in the ratio of 49.5:49.5:1 (v/v/v); saturated with 100 mM TRIS pH 8.0; contains ~0.1% 8-hydroxyquinoline

Other Notes

Large- and small-scale phenol extractions[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7761977617516726
grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid (Two-layer)

density

1.29 g/mL at 20 °C

density

1.12 g/mL at 20 °C

density

1.28 g/mL at 20 °C

density

-

impurities

DNases, none detected, phosphatases, none detected, RNases, none detected, proteases, none detected

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

impurities

-

pH

7.7-8.3(H2O-phase, after extraction with H2O, 1:1)

pH

4-5(H2O-phase, after extraction with H2O, 1:1)

pH

7.2-8.3(H2O-phase, after extraction with H2O, 1:1)

pH

-


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 3

target_organs

Central nervous system, Liver,Kidney, Nervous system,Kidney,Liver,Skin

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

1 of 1 reviewers took part in a promotion

Active Filters

    • Review 1
    • Votes 0
    1 out of 5 stars.

    Phenol-Chloroform degraded

    The Phenol-Chloroform degraded. We called your company and has not receive any replacement yet..

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for your review. We are sorry to hear that your experience did not match your product expectations. We encourage customers who experience problems with our products to call or email us for additional technical support. Please visit https://www.sigmaaldrich.com/US/en/support/customer-support to submit a Product Technical Inquiry.