Skip to Content
MilliporeSigma

8.18969

Isoamyl alcohol

for synthesis

Synonym(s):

3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHCH2CH2OH
CAS Number:
Molecular Weight:
88.15
UNSPSC Code:
12352104
NACRES:
NA.21
EC Index Number:
204-633-5
MDL number:
Beilstein/REAXYS Number:
1718835
Assay:
≥98% (GC)
Grade:
synthesis grade
Bp:
130 °C (lit.)
Vapor pressure:
2 mmHg ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 20 °C)

assay

≥98% (GC)

form

liquid

autoignition temp.

644 °F

potency

>5000 mg/kg LD50, oral (Rat), >3000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-9 %, 100 °F

dilution

(for synthesis)

refractive index

n20/D 1.406 (lit.)

pH

7 (20 °C, 25 g/L in H2O)

kinematic viscosity

5.32 cSt(20 °C)

bp

130 °C (lit.)

mp

−117 °C (lit.)

transition temp

flash point 43.5 °C

solubility

25 g/L

density

0.809 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)CCO

InChI

1S/C5H12O/c1-5(2)3-4-6/h5-6H,3-4H2,1-2H3

InChI key

PHTQWCKDNZKARW-UHFFFAOYSA-N

Features and Benefits

Saturation Concentration in Air: 11 g/m3 (20 °C) Air

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.808 - 0.809
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Isoamyl alcohol


Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

3 - Flammable liquids

Flash Point (°F)

110.3 °F - closed cup

Flash Point (°C)

43.5 °C - closed cup

Signal Word

Danger



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library