Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

5010-OP

Millipore

Guanidine Hydrochloride

OmniPur® Grade, >= 99.5%, Denaturant used for purification of nucleic acids and polypeptides from cell lysates

Synonym(s):

Guanidine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.28

Quality Level

form

crystalline powder (granular)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

mp

180-185 °C (lit.)

density

1.3 g/cm3 (lit.)

foreign activity

DNAse, none detected
Phosphatase, none detected
Protease, none detected
RNAse, none detected

shipped in

ambient

storage temp.

10-30°C

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description



Denaturant used for purification of nucleic acids and polypeptides from cell lysates. Inhibits RNase and protease activity. Molecular Biology grade.br
A260 (0.1 M, water): ≤0.05 AU
A280 (0.1 M, water): ≤0.01 AU
Purity by Assay: ≥99.5%
DNase: None detected
Heavy metals (as Pb): ≤0.001%
Phosphatase: None detected
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service