Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50937

Sigma-Aldrich

Guanidine hydrochloride solution

BioUltra, ~8 M in H2O

Synonym(s):

Aminoformamidine hydrochloride, Guanidinium chloride, Guanidium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N3 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

form

liquid

concentration

~8 M in H2O

impurities

insoluble matter, passes filter test

refractive index

n20/D 1.465

pH

4.5-7.5 (25 °C)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤5 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤150 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.040
λ: 280 nm Amax: 0.025

SMILES string

Cl.NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Guanidine hydrochloride solution has been used as a fixation buffer in genomic DNA extraction for genomic analysis.

Biochem/physiol Actions

Guanidine‐HCl (Gnd-HCl), an efficient chaotropic agent, is broadly used in RNA isolation, globular protein denaturation, and protein refolding studies as a denaturant. It is an appropriate sample buffer for the quick and effective elution of affinity-purified protein complexes and the subsequent protein digestion step. Gnd-HCl is not preferred in proteomics experiments due to its hindrance with trypsin activity at low molar concentrations and tendency to precipitate when combined with sodium dodecyl sulfate (SDS). GnHCl is commonly utilized to extract proteins in extracellular matrix (ECM)‐rich tissues including bone, cartilage, ligament, and tendon.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Class III Peroxidases in Response to Multiple Abiotic Stresses in Arabidopsis thaliana Pyrenean Populations
Eljebbawi A, et al.
International Journal of Molecular Sciences (2022)
Yalda Ashraf Kharaz et al.
Proteomics, 17(13-14) (2017-05-27)
Exploring the tendon proteome is a challenging but important task for understanding the mechanisms of physiological/pathological processes during ageing and disease and for the development of new treatments. Several extraction methods have been utilised for tendon mass spectrometry, however different
Jon W Poulsen et al.
Journal of proteome research, 12(2), 1020-1030 (2012-11-29)
Protein digestion is an integral part of the "shotgun" proteomics approach and commonly requires overnight incubation prior to mass spectrometry analysis. Quadruplicate "shotgun" proteomic analysis of whole yeast lysate demonstrated that Guanidine-Hydrochloride (Gnd-HCl) protein digestion can be optimally completed within
Caitriona McKeever et al.
Journal of medicinal chemistry, 56(3), 700-711 (2013-01-11)
Considering the strong DNA minor groove binding observed for our previous series of diaromatic symmetric and asymmetric guanidinium and 2-aminoimidazolinium derivatives, we report now the synthesis of new aminoalkyl derivatives of diaromatic guanidines with potential as DNA minor groove binders
Allan Joaquim Lamontanara et al.
Nature communications, 5, 5470-5470 (2014-11-18)
The activity of protein kinases is regulated by multiple molecular mechanisms, and their disruption is a common driver of oncogenesis. A central and almost universal control element of protein kinase activity is the activation loop that utilizes both conformation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service