Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369075

Millipore

Guanidine Hydrochloride

ULTROL® Grade, ≥99% (titration), protein denaturant for purification of proteins, nucleic acids, and mRNA

Synonym(s):

Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH5N3 · xHCl
CAS Number:
Molecular Weight:
59.07 (free base basis)
MDL number:
UNSPSC Code:
12352107
NACRES:
NA.25

Quality Level

Assay

≥99% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

ethanol: soluble
water: soluble

shipped in

ambient

storage temp.

15-25°C

SMILES string

[Cl-].[N+H3]\C(=N\[H])\N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A powerful protein denaturant that is widely used in the purification of proteins, nucleic acids, and mRNA. It is used in a variety of applications, including sequencing, molecular weight determinations, and as a solubilizing agent for electrophoresis.
Absorbance (6.0 M, H2O): <0.2 at 230 nm; <0.03 at 260 nm.

Application


  • Revealing the Roles of Guanidine Hydrochloride Ionic Liquid in Ion Inhibition and Defects Passivation for Efficient and Stable Perovskites Solar Cells: Discusses how guanidine hydrochloride ionic liquid enhances the efficiency and stability of perovskite solar cells by ion inhibition and defect passivation (Saeed A et al., 2024).

  • N-Fluorosulfonyl Guanidine: An Entry to N-Guanyl Sulfamides and Sulfamates: Investigates the use of N-fluorosulfonyl guanidine for synthesizing N-guanyl sulfamides and sulfamates, showcasing its potential in chemical synthesis and material science (Wang W et al., 2024).

  • Efficient extraction of lignin from moso bamboo by microwave-assisted ternary deep eutectic solvent pretreatment for enhanced enzymatic hydrolysis: Explores the role of guanidine hydrochloride in enhancing the enzymatic hydrolysis of lignin extracted from moso bamboo using a microwave-assisted deep eutectic solvent method (Feng Y et al., 2024).

  • Computational Enzyme Redesign Enhances Tolerance to Denaturants for Peptide C-Terminal Amidation: Details how computational redesign of enzymes, involving guanidine hydrochloride, can enhance their tolerance to denaturants, improving the efficiency of peptide C-terminal amidation processes (Zhu T et al., 2024).

  • Parasite Burden of Trypanosoma cruzi in Whole Blood and Buffy Coat Determined by Real-Time PCR in Individuals with Chronic Chagas Disease: Describes the use of guanidine hydrochloride in real-time PCR processes to quantify the parasite burden in blood samples from individuals with Chagas disease, demonstrating its utility in enhancing diagnostic assays (Liempi D et al., 2024).

Warning

Toxicity: Harmful (C)

Other Notes

Bastiras, S., and Wallace, J.C. 1992. Biochemistry31, 9304.
Plaza del Pino, I.M., et al. 1992. Biochemistry31, 11196.
Steinman, H., et al. 1974. J. Biol. Chem.249, 7326.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTROL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service