Skip to Content
MilliporeSigma
All Photos(1)

Documents

48-655MAG

Millipore

MILLIPLEX® Protein Translation Magnetic Bead 6-Plex Kit - Cell Signaling Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human, rat, mouse

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Protein synthesis is required for cellular growth and development. Normal cells grow in a controlled manner in response to environmental and developmental cues. However, cancer cells can reprogram cellular metabolism, favoring uncontrolled growth and survival. This can be achieved by altering signaling pathways that control cellular processes such as protein synthesis.The phosphorylation state of proteins involved in translation initiation is a limiting factor that regulates the formation or activity of translational complexes. In cancer cells, hyper-activated signaling pathways influence translation, allowing uncontrolled growth and survival. In addition, several components of translation initiation have been found to be mutated, post-translationally modified, or differentially expressed, and thus have been shown to act as oncogenes.Translational alterations can increase the overall rate of protein synthesis as well as activate regulatory mechanisms leading to the translation of specific messenger RNAs for proteins that promote cancer progression and survival.Each MILLIPLEX® cell signaling kit includes:• Stimulated and unstimulated cell lysates provided to qualify assay performance• Premixed magnetic beads to capture analytes of interest• Optimized detection antibody cocktails designed to yield consistent analyte profiles within a panelThe MILLIPLEX® Protein Translation 6-plex Magnetic Bead kit is used to detect changes in phosphorylated eIF2a (Ser51), eIF-4B (Ser422), eIF-4E (Ser209), eIF-4G (Ser1108) and 4E-BP1 (Thr37/46), as well as total protein levels of 4E-BP1 in cell lysates using the Luminex® system. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures. Each kit has sufficient reagents for one 96-well plate assay.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 2 (provided).1 - 25 μg cell lysate/well (recommended starting concentration is 40 to 1,000 μg protein/mL).Analytes available:eIF-4G (Ser1108);elF-4E (Ser209);elF-4B (Ser422);elF-2a (Ser51);4E-BP1 (Total);4E-BP1 (Thr37/46)

Packaging

96-well plate

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service