Skip to Content
MilliporeSigma
All Photos(1)

Documents

48-682MAG

Millipore

MILLIPLEX® Bcl-2 Family Apoptosis Panel 1 Magnetic Bead 6-plex Kit - Cell Signaling Multiplex Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Apoptosis is an essential physiological process that plays a crucial role in normal development and homeostatic mechanisms of multicellular organisms. At least two major apoptotic pathways have been described in mammalian cells: the intrinsic pathway via Caspase-9 activation, in which death arises from mitochondrial dysfunction, and the extrinsic pathway via Caspase-8 activation, in which death is initiated from the activation of cell surface receptors. Bcl-2 family members play an important role in the intrinsic apoptotic pathway and includes over 17 members. Interactions of these Bcl-2 family members function as a ‘life/death switch′ that integrates diverse inter- and intracellular cues to determine whether or not the stress apoptosis pathway should be activated. For example, Bcl-2 and its closely related anti-apoptotic proteins, Bcl-xL and Mcl-1, can sequester their pro-apoptotic counterparts (BAD, BIM, NOXA and PUMA) and thereby inhibit the initiation steps of apoptosis..The MILLIPLEX® MAP 6-plex Bcl-2 Family Apoptosis Panel 1 Magnetic Bead Kit (Cat. No. 48-682MAG) is a premixed kit to be used for the simultaneous quantification of the following 6 analytes:pBAD (Ser112)BAD (total)BIM (total)BAX (total)Bcl-xL/BADMcl-1/BIM

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enable the simultaneous relative quantitation of multiple phosphorylation and total pathway proteins in tissue and cell lysate samples. Compare Multiplexing results to those of Western blotting.An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 1 (provided).The suggested working range of protein concentration for the assay is 1 to 25 μg of total protein/well (25 μL/well at 40 to 1,000 μg/mL).Analytes available:BAD (Ser112);BAD (Total);BIM;BAX;Bcl-xL/BAD;Mcl-1/BIM

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

Multiplex assays simplify cancer research by measuring multiple biomarkers simultaneously for various signaling pathways.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service