Skip to Content
MilliporeSigma
All Photos(1)

Documents

48-618MAG

Millipore

MILLIPLEX® Total/Phospho Akt - Cell Signaling Multiplex Assay

Synonym(s):

Akt Magnetic Bead Kit, Cell Signaling Multiplex Kit, Total/Phospho Akt Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

species reactivity

human, mouse, rat

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

The MILLIPLEX® Total/Phospho Akt (48-618MAG) and Total/Phospho ERK (Catalog No. 48-619) Magnetic bead kits have been developed for the simultaneous detection of phosphorylated Akt (Ser473) and total Akt or phosphorylated ERK1/2 (Thr185/Tyr187) and total ERK in a single well using the Luminex system. These detection assays are rapid, convenient alternatives to Western Blotting and immunoprecipitation procedures for the analysis of cell lysate samples.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

Intracellular Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous relative quantitation of multiple phosphorylation or total pathway proteins in tissue and cell lysate samples. An overnight (4°C) incubation is recommended for best results.This assay requires 25 μL diluted cell lysate per well.This kit must be run using Assay Buffer 2 (provided).1 - 25 μg cell lysate/well (recommended starting concentration is 40 to 1,000 μg protein/mL)Analytes available:Akt (Total)Akt (Ser473)

Packaging

96-well plate

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service