Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

479775

Sigma-Aldrich

α-Naphthyl Acid Phosphate monosodium salt

≥98% (HPLC), solid, broad-spectrum protein phosphatase inhibitor, Calbiochem®

Synonym(s):

α-Naphthyl Acid Phosphate, Monosodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8NaO4P
CAS Number:
Molecular Weight:
246.13
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Product Name

α-Naphthyl Acid Phosphate, Monosodium Salt, α-Naphthyl Acid Phosphate, Monosodium Salt - CAS 81012-89-7, is a broad-spectrum protein phosphatase inhibitor.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

<0.01% α-naphthol

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[P](=O)([O-])([O-])Oc1c2c(ccc1)cccc2

InChI

1S/C10H9O4P/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H2,11,12,13)/p-2

InChI key

YNXICDMQCQPQEW-UHFFFAOYSA-L

General description

Broad-spectrum protein phosphatase inhibitor.

Biochem/physiol Actions

Cell permeable: no
Primary Target
protein phosphatase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Davidson, M.M., and Haslam, R.J. 1994. Biochem. J. 301, 41.
Wolniak, S.M., and Larsen, P.M. 1992. J. Cell Sci.102, 691.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service