Skip to Content
MilliporeSigma

N7375

2-Naphthyl phosphate sodium salt

≥98% (TLC), powder

Synonym(s):

β-Naphthyl phosphate monosodium salt, 2-Naphthyl phosphate monosodium salt, mono-Sodium 2-naphthyl phosphate sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8NaO4P
CAS Number:
Molecular Weight:
246.13
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
238-452-8
MDL number:
Beilstein/REAXYS Number:
1969493
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Naphthyl phosphate sodium salt,

assay

≥98% (TLC)

Quality Segment

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].OP([O-])(=O)Oc1ccc2ccccc2c1

InChI

1S/C10H9O4P.Na/c11-15(12,13)14-10-6-5-8-3-1-2-4-9(8)7-10;/h1-7H,(H2,11,12,13);/q;+1/p-1

InChI key

GHMHDIFFBHLXTJ-UHFFFAOYSA-M



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library