Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N5602

Sigma-Aldrich

1-Naphthyl phosphate

99%

Synonym(s):

α-Naphthol phosphate, α-Naphthyl acid phosphate, α-Naphthyl phosphate, 1-Naphthyl dihydrogen phosphate, 1-Naphthyl phosphate (7CI), Naphthol 1-phosphate, Naphthyl dihydrogen phosphate, Phosphoric acid α-naphthyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9O4P
CAS Number:
Molecular Weight:
224.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

mp

157-159 °C (lit.)

SMILES string

OP(O)(=O)Oc1cccc2ccccc12

InChI

1S/C10H9O4P/c11-15(12,13)14-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,(H2,11,12,13)

InChI key

YNXICDMQCQPQEW-UHFFFAOYSA-N

Gene Information

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Brici et al.
Development (Cambridge, England), 144(14), 2606-2617 (2017-06-18)
Germ cell development involves major reprogramming of the epigenome to prime the zygote for totipotency. Histone 3 lysine 4 (H3K4) methylations are universal epigenetic marks mediated in mammals by six H3K4 methyltransferases related to fly Trithorax, including two yeast Set1
Giulia Selvolini et al.
Sensors (Basel, Switzerland), 18(7) (2018-06-27)
In this work, we propose an electrochemical DNA aptasensor for the detection of profenofos, an organophosphorus pesticide, based on a competitive format and disposable graphite screen-printed electrodes (GSPEs). A thiol-tethered DNA capture probe, which results to be complementary to the
Zhanna Lipatova et al.
PLoS genetics, 16(12), e1009255-e1009255 (2020-12-05)
Thirty percent of all cellular proteins are inserted into the endoplasmic reticulum (ER), which spans throughout the cytoplasm. Two well-established stress-induced pathways ensure quality control (QC) at the ER: ER-phagy and ER-associated degradation (ERAD), which shuttle cargo for degradation to
Eva Vargas et al.
Biosensors & bioelectronics, 167, 112512-112512 (2020-09-03)
Here we describe the development of a dual electrochemical immunosensor microchip for simultaneous detection of insulin (I) and cortisol (C) biomarkers that can enhance the ability to improve glucose regulation using automated insulin delivery. The successful realization of the simultaneous
Gözde Aydoğdu Tığ et al.
Talanta, 210, 120666-120666 (2020-01-29)
In this work, we reported an electrochemical aptasensor based on the poly-3-amino-1,2,4-triazole-5-thiol/graphene oxide composite (P(ATT)-GO) and gold nanoparticles (AuNPs) modified graphite screen-printed electrode (GSPE) (GSPE/P(ATT)-GO/AuNPs) for determination of lipocalin-2 (LCN2) (neutrophil gelatinase-associated lipocalin). A sandwich based strategy was utilized to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service