Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

444252

Millipore

MMP-9/MMP-13 Inhibitor I

The MMP-9/MMP-13 Inhibitor I, also referenced under CAS 204140-01-2, controls the biological activity of MMP-9/MMP-13. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-9/MMP-13 Inhibitor I, N-Hydroxy-1-(4-methoxyphenyl)sulfonyl-4-(4-biphenylcarbonyl)piperazine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25N3O6S
CAS Number:
Molecular Weight:
495.55
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

methanol: 1 mg/mL
DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable and highly, piperazine-based potent inhibitor of MMP-9 (IC50 = 900 pM) and MMP-13 (IC50 = 900 pM). Inhibits MMP-1 and MMP-3 at much higher concentrations (IC50 = 43 nM and 23 nM, respectively). Also acts as an inhibitor of MMP-7 (IC50 = 930 nM).
A piperazine-based, cell-permeable, and highly potent inhibitor of MMP-9 (IC50 = 900 pM) and MMP-13 (IC50 = 900 pM). Inhibits MMP-1 and MMP-3 at much higher concentrations (IC50 = 43 nM and 23 nM, respectively). Also acts as an inhibitor of MMP-7 (IC50 = 930 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-9
Product does not compete with ATP.
Reversible: no
Target IC50: 900 pM against both MMP-9 and MMP-13

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cheng, M., et al. 2000. J. Med. Chem.43, 369.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hao Huang et al.
Cell reports, 36(2), 109363-109363 (2021-07-15)
Although activating mutations of the anaplastic lymphoma kinase (ALK) membrane receptor occur in ∼10% of neuroblastoma (NB) tumors, the role of the wild-type (WT) receptor, which is aberrantly expressed in most non-mutated cases, is unclear. Both WT and mutant proteins
Benjamin Seyer et al.
Journal of neurochemistry, 153(4), 485-494 (2019-09-27)
Ethyl2-acetylamino-7-hydroxy-4-pyridin-3-yl-4H-chromene-3-carboxylate (HFI-419), the benzopyran-based inhibitor of insulin-regulated aminopeptidase (IRAP), has previously been shown to improve spatial working and recognition memory in rodents. However, the mechanism of its cognitive-enhancing effect remains unknown. There is a close correlation between dendritic spine density

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service