Skip to Content
MilliporeSigma

Skip To

SML0583

Ferrostatin-1

≥95% (HPLC), powder, non-apoptotic cell death inhibitor

Synonym(s):

3-Amino-4-cyclohexylaminobenzoic acid ethyl ester;

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H22N2O2
CAS Number:
Molecular Weight:
262.35
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ferrostatin-1, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

color

, faint purple to brown

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(C2CCCCC2)c1c(cc(cc1)C(=O)OCC)N

InChI

1S/C15H22N2O2/c1-2-19-15(18)11-8-9-14(13(16)10-11)17-12-6-4-3-5-7-12/h8-10,12,17H,2-7,16H2,1H3

InChI key

UJHBVMHOBZBWMX-UHFFFAOYSA-N

Application

Ferrostatin-1 has been used to determine its inhibitory effect on the cell death profiles.

Biochem/physiol Actions

Ferrostatin-1 is a potent inhibitor of non-apoptotic cell death induced by erastin call ferroptosis.
Ferrostatin-1 is a potent inhibitor of non-apoptotic cell death induced by erastin call ferroptosis. Ferrostatin-1 prevents ferroptopic cell death in cancer cells and glutamate-induced toxicity in organotypic rat brain slices. It appears that Ferrostatin-1 controls lipid soluble ROS.
Ferrostatin-1 is an active radical-trapping antioxidant that traps peroxyl radicals, and thus serves as a potential inhibitor of ferroptosis. Ferroptosis is considered as regulated necrosis, which differs from apoptosis and autophagy. Ferrostatin-1 is known to show protective effects against Huntington′s disease and also prevents neurotoxicity induced by glutamate in cellular models.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1925SML1656SML0424
description

≥95% (HPLC)

description

≥95% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

paste

form

powder

assay

≥95% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

H2O: 10 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 10 mg/mL (clear solution)

color

, faint purple to brown

color

white to beige

color

white to beige

color

white to beige


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. DMSOで溶解後の保存は冷蔵保存ですか?それとも冷凍保存でしょうか?

    1 answer
    1. Following reconstitution, stock solutions are stable up to 6 months when stored frozen in aliquots at -20°C.

      Helpful?

  2. What is the protocol for diluting SML0583 (5 mg), which is said to be soluble in DMSO? Additionally, what kind of stock solution can I prepare for long-term storage and where to store it after adding DMSO.

    1 answer
    1. The solubility of Ferrostatin-1 is approximately 10 mg/ml in DMSO. The detailed solution stability information is not available; therefore, it is recommended to prepare a fresh solution for each application.

      Helpful?

Reviews

No rating value

Active Filters