Skip to Content
MilliporeSigma
All Photos(1)

Documents

438185

Sigma-Aldrich

Lovastatin

Lovastatin, CAS 75330-75-5, is an anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase.

Synonym(s):

Lovastatin, Mevinolin, MK-803, L-Type Calcium Channel Blocker IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

ethanol: 25 mg/mL
DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

PCZOHLXUXFIOCF-BXMDZJJMSA-N

General description

An anti-hypercholesterolemic agent and an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase that blocks a series of biological events including: activation of p21ras by insulin in 3T3-L1 fibroblasts and 3T3-L1 adipocytes, farnesylation of p21ras, which decreases the pool of intracellular Ras available for subsequent activation by growth factors (including insulin), and N-ras-induced neuronal differentiation. Causes cell cycle arrest in the late G1 phase. Shown to inhibit the stimulation of MAP kinase by insulin in HIRcB cells and block the transcription of the type-I SCR gene in THP-1-derived macrophages. Also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF. This product requires activation for use in cell synchronization studies and in cell-free assay systems for HMG-CoA studies.
An anti-hypercholesterolemic agent that inhibits the activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase. Has been shown to block a series of biological events including: the activation of p21ras by insulin in 3T3-L1 fibroblasts and 3T3-L1 adipocytes; the farnesylation of p21ras, which decreases the pool of intracellular Ras available for subsequent activation by growth factors (including insulin); and N-ras-induced neuronal differentiation. Causes cell cycle arrest in the late G1 phase through inhibition of proteasome. Has recently been shown to inhibit the stimulation of MAP kinase by insulin in HIRcB cells and to block the transcription of type-I SCR gene in THP-1-derived macrophages. Also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF. This product requires activation for use in cell synchronization studies and in cell-free assay systems for HMG-CoA studies.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Activity of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution aliquot, flush with an inert gas, and freeze (-20°C). Stock solutions, in high quality DMSO or ethanol are stable for up to 1 month at -20°C.

Other Notes

Rao, S., et al. 1999. Proc. Natl. Acad. Sci. USA96, 7197.
Carel, K., et al. 1996. J. Biol. Chem.271, 30625.
McGuire, T.F., et al. 1996. J. Biol. Chem. 271, 27402.
Umetani, N., et al. 1996. Biochim. Biophys. Acta1303, 199.
Xu, X.Q., et al. 1996. Arch. Biochem. Biophys. 326, 233.
Reusch, J.E.-B., et al. 1995. J. Biol. Chem.270, 2036.
Jakobisiak, M., et al. 1991. Proc. Natl. Acad.Sci. USA88, 3628.
Keyomarsi, K., et al. 1991. Cancer Res.51, 3602.
Mendola, C.E., and Backer, J.M. 1990. Cell Growth Differ. 1, 499.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marielle Cochet et al.
Veterinary research, 55(1), 32-32 (2024-03-17)
Outbreaks of West Nile virus (WNV) occur periodically, affecting both human and equine populations. There are no vaccines for humans, and those commercialised for horses do not have sufficient coverage. Specific antiviral treatments do not exist. Many drug discovery studies
Martha M Monick et al.
Journal of immunology (Baltimore, Md. : 1950), 171(5), 2625-2630 (2003-08-21)
These studies demonstrate that treatment of macrophages with lovastatin, a cholesterol-lowering drug that blocks farnesylation and geranylgeranylation of target proteins, increases LPS-induced TNF-alpha production. This is reversed by the addition of mevalonate, which bypasses the lovastatin block. Examination of membrane
Woo-Jin Lim et al.
Cells, 10(9) (2021-09-29)
Retrospective observational studies have reported that statins improve clinical outcomes in patients previously treated with programmed cell death protein 1 (PD-1)-targeting monoclonal antibodies for malignant pleural mesothelioma (MPM) and advanced non-small cell lung cancer (NSCLC). In multiple mouse cancer models
Kei K Ito et al.
The Journal of cell biology, 220(3) (2021-01-26)
Centrioles duplicate in interphase only once per cell cycle. Newly formed centrioles remain associated with their mother centrioles. The two centrioles disengage at the end of mitosis, which licenses centriole duplication in the next cell cycle. Therefore, timely centriole disengagement
David W Sanders et al.
eLife, 10 (2021-04-24)
Many enveloped viruses induce multinucleated cells (syncytia), reflective of membrane fusion events caused by the same machinery that underlies viral entry. These syncytia are thought to facilitate replication and evasion of the host immune response. Here, we report that co-culture

Articles

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Discover critical characteristics to consider when working with enzyme inhibitors, such as cell permeability, the prozone effect, and Lipinski’s rule of 5.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service