Skip to Content
MilliporeSigma
All Photos(1)

Documents

438186

Sigma-Aldrich

Lovastatin sodium salt

≥95% (HPLC), solid, L-type calcium channel blocker, Calbiochem®

Synonym(s):

Lovastatin, Sodium Salt, L-Type Calcium Channel Blocker IV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H37O6 · Na
Molecular Weight:
444.54
UNSPSC Code:
51111800
NACRES:
NA.77

product name

Lovastatin, Sodium Salt, Carboxylate form of Lovastatin that is active in whole cells and cell-free assays.

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

off-white

solubility

DMSO: 100 mg/mL
ethanol: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Carboxylate form of Lovastatin (Cat. No. 438185) that is active in whole cells and cell-free assays.
Carboxylate form of Lovastatin (Cat. No. 438185) that is active in whole cells and cell-free assays. Lovastatin is reported to be an anti-hypercholesterolemic agent that acts as an inhibitor of 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase and blocks a series of biological events including activation of p21ras by insulin in 3T3-L1 fibroblasts and 3T3-L1 adipocytes, farnesylation of p21ras, which decreases the pool of intracellular Ras available for subsequent activation by growth factors (including insulin), and N-ras-induced neuronal differentiation. Causes cell cycle arrest in the late G1 phase. Shown to inhibit the stimulation of MAP kinase by insulin in HIRcB cells and block the transcription of the type-I SCR gene in THP-1-derived macrophages. Also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) reductase
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot, flush with an inert gas, and freeze (-20°C). Stock solutions in high quality ethanol are stable for up to 1 month at -20°C.

Other Notes

Rao, S., et al. 1999. Proc. Natl. Acad. Sci. USA96, 7197.
Carel, K., et al. 1996. J. Biol. Chem.271, 30625.
McGuire, T.F., et al. 1996. J. Biol. Chem.271, 27402.
Umetani, N., et al. 1996. Biochim. Biophys. Acta1303, 199.
Xu, X.Q., et al. 1996. Arch. Biochem. Biophys.326, 233.
Reusch, J.E.-B., et al. 1995. J. Biol. Chem.270, 2036.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service