Skip to Content
MilliporeSigma
All Photos(1)

Documents

344095-M

Millipore

Fluvastatin, Sodium Salt

A synthetic HMG-CoA reductase inhibitor (IC₅₀ = 40-100 nM for human liver microsomes) that acts as an anti-hypercholesterolemic agent.

Synonym(s):

Fluvastatin, Sodium Salt, (±)-(3Rʹ,5Sʹ,6E)-7-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]-3,5-dihydroxy-6-heptenoate, sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

water: 30 mg/mL
DMSO: soluble
methanol: soluble

storage temp.

−20°C

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

General description

A synthetic HMG-CoA reductase inhibitor (IC50 = 40-100 nM for human liver microsomes) that acts as an anti-hypercholesterolemic agent. Decreases the basal MMP-1 levels in culture media of endothelial cells in a time- and dose-dependent manner. Inhibits the formation of TBA-reactive substances in Fe(II)-supported peroxidation of liposomes (IC50 = 12 µM). Also available as a 10 mM solution in H2O(Cat. No. 344096).
A synthetic, competitive inhibitor of HMG-CoA reductase that acts as anti-hypercholesterolemic agent. Unlike Lovastatin (Cat. No. 438185), Mevastatin (Cat. No. 474700), and Simvastatin (Cat. No. 567020), it does not require activation prior to use for in vitro studies. Reduces the basal MMP-1 levels in culture media of endothelial cells in a time- and dose-dependent manner. Fluvastatin is metabolized in the liver, primarily via hydroxylation of the indole ring at the 5- and 6-positions. N-Dealky-lation and b-oxidation of the side-chain also occurs. The hydroxy metabolites are also reported to have some pharmacologic activity.

Biochem/physiol Actions

Primary Target
HMG-CoA reductase
Target IC50: 40-100 nM against HMG-CoA reductase in human liver microsomes

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Yamamoto, A., et al. 2001. J. Pharm. Pharmacol.53, 227.
Dansette, P.M., et al. 2000. Exp. Toxicol. Pathol.52, 145.
Ikeda, U., et al. 2000. Hypertension36, 325.
Levy, R.I., et al. 1993. Circulation87 (4 Suppl.), III45.
Tse, F.L.S. et al. 1992. J. Clin. Pharmacol.32, 630.
Yuan, J., et al. 1991. Atherosclerosis87, 147.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service