Skip to Content
MilliporeSigma

444968

PD 0325901

≥95% (HPLC), liquid, MEK1/2 inhibitor, Calbiochem®

Synonym(s):

InSolution MEK1/2 Inhibitor III, PD0325901, PD 0325901, N-((2R)-2,3-Dihyroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)-benzamide, PD325901

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H14F3IN2O4
CAS Number:
Molecular Weight:
482.19
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
liquid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

MEK1/2 Inhibitor III, PD0325901, InSolution, ≥95%

Quality Segment

assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

shipped in

ambient

storage temp.

−20°C

SMILES string

Ic1cc(c(cc1)Nc2c(c(ccc2C(=O)NOC[C@H](O)CO)F)F)F

InChI

1S/C16H14F3IN2O4/c17-11-3-2-10(16(25)22-26-7-9(24)6-23)15(14(11)19)21-13-4-1-8(20)5-12(13)18/h1-5,9,21,23-24H,6-7H2,(H,22,25)/t9-/m1/s1

InChI key

SUDAHWBOROXANE-SECBINFHSA-N

General description

PD-0325901 is a highly effective oral small-molecule inhibitor that targets both mitogen-activated protein kinase kinase 1 (MEK1) and MEK2. It is a selective, non-ATP-competitiveinhibitor of these two MEK isoforms. This cell-permeable benzamide compound is a selective non-competitive inhibitor of MAPKK/MKK, effectively blocking cellular Erk1/2 phosphorylation. In serum-starved HeLa cells, it achieves over 90% inhibition after serum stimulation, while in C26 cells, it reduces phosphorylation by 50% at inhibitor concentrations of 25 nM and 0.33 nM, respectively. The compound exhibits minimal to no effect on Erk1/2 activity ora panel of 66 other kinases, even at concentrations as high as 10 µM in cell-free kinase assays. PD-0325901 has demonstrated oral bioavailability inmice, successfully suppressing both tumor growth and Erk phosphorylation in BRAF(V600E)-expressing SKMEL28 cell-derived tumors in vivo.

Application

PD 0325901 has been used as:·

  • a supplement in induced pluripotent stem cells (iPSC) medium to culture MEF feeder layers with reprogrammed cells
  • mitogen-activated protein kinase kinase1/2 (MEK1/2) inhibitor to treat the cells to identify G1 phase cells that had crossed the restriction point in the cell cycle
  • Mek1/2 inhibitor to block the Ras-Erk1/2 signaling pathwayto investigate the role of fibroblast growth factor (Fgf) signaling in the specification of endoderm progenitors during embryonic development

Packaging

Packaged under inert gas

Physical form

A 50 mM (5 mg/207 µl) solution of MEK1/2 Inhibitor III, PD0325901 (Cat. No. 444966 ) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Regulatory Review (Z)


Still not finding the right product?

Explore all of our products under MEK1/2 Inhibitor III, PD0325901


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

188.6 °F - closed cup - (refers to pure substance)

flash_point_c

87 °C - closed cup - (refers to pure substance)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library