Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

317240

Sigma-Aldrich

Metformin

≥97% (titration), solid, AMPK signaling activator, Calbiochem®

Synonym(s):

Metformin, 1,1-Dimethylbiguanide, Hydrochloride, AMPK Signaling Activator VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11N5 · xHCl
CAS Number:
Molecular Weight:
129.16 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Metformin, An antihyperglycemic agent that lowers blood glucose levels without stimulating insulin secretion, promoting weight gain or causing hypoglycemia.

Quality Level

description

Merck USA index - 14, 5938

Assay

≥97% (titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 50 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

Cl.N(C)(C)C(=N\C(=N\[H])\N)N

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H

InChI key

OETHQSJEHLVLGH-UHFFFAOYSA-N

General description

An anti-hyperglycemic agent that acts by lowering the hepatic output of glucose. Does not stimulate insulin secretion, promote weight gain, or cause hypoglycemia. Reported to stimulate AMP-activated protein kinase (AMPK), leading to the reduction in acetyl-CoA carboxylase (ACC) activity and induction of fatty acid oxidation.
An antihyperglycemic agent that lowers blood glucose levels without stimulating insulin secretion, promoting weight gain or causing hypoglycemia. Reported to stimulate AMP-activated protein kinase (AMPK) leading to the reduction in acetyl-CoA carboxylase (ACC) activity and induction of fatty acid oxidation.

Biochem/physiol Actions

Cell permeable: no
Primary Target
AMP-activated protein kinase (AMPK)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cleasby, M.E., et al. 2004. Diabetes53, 3258.
Klein, J., et al. 2004. J. Endocrinol.183, 299.
Zhou, G., et al. 2001. J. Clin. Invest.108, 1167.
Lin, H.Z., et al. 2000. Nat. Med.6, 998.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas M Wiggenhauser et al.
Diabetes, 69(5), 1020-1031 (2020-03-07)
Progression from the initial vascular response upon hyperglycemia to a proliferative stage with neovacularizations is the hallmark of proliferative diabetic retinopathy. Here, we report on the novel diabetic pdx1 -/- zebrafish mutant as a model for diabetic retinopathy that lacks
Martina Poteti et al.
Cancers, 13(17) (2021-09-11)
This study was directed to characterize the role of glutamine in the modulation of the response of chronic myeloid leukemia (CML) cells to low oxygen, a main condition of hematopoietic stem cell niches of bone marrow. Cells were incubated in

Questions

1–2 of 2 Questions  
  1. What is the solubility of Metformin (Cat#317240) in DMSO?

    1 answer
    1. The substance dissolves in DMSO at concentrations of up to 8 mg/ml.

      Helpful?

  2. is metformin accociated with paracitic worms?

    1 answer
    1. There have been studies published regarding the efficacy of Metformin against various parasites. Please see the link below to review one such study of Echinococcisis. Note that this product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC5321462/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service