Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420321

Sigma-Aldrich

KT5823

Highly specific, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase G (Ki = 234 nM).

Synonym(s):

KT5823, PKG Inhibitor VII

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H25N3O5
CAS Number:
Molecular Weight:
495.53
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 1 mg/mL
DMF: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[n]21c3c4[n](c6c(c4c7c(c3c8c2cccc8)CN(C7=O)C)cccc6)[C@@H]5O[C@]1([C@@](C5)(OC)C(=O)OC)C

InChI

1S/C29H25N3O5/c1-28-29(36-4,27(34)35-3)13-20(37-28)31-18-11-7-5-9-15(18)22-23-17(14-30(2)26(23)33)21-16-10-6-8-12-19(16)32(28)25(21)24(22)31/h5-12,20H,13-14H2,1-4H3/t20-,28+,29+/m1/s1

InChI key

QTYMDECKVKSGSM-YMUMJAELSA-N

General description

Highly specific cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase G (Ki = 234 nM). Inhibits protein kinase C (Ki = 4.0 µM) and protein kinase A (Ki >10.0 µM) at higher concentrations.
Highly specific, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase G (Ki = 234 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKG
Product competes with ATP.
Reversible: yes
Target Ki: 234 nM against protein kinase G

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Grider, J.R. 1993. Am. J. Physiol.264, G334.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyu Zhang et al.
Cells, 11(23) (2022-12-12)
Spiral ganglion neurons (SGNs) are important for hearing, and their peripheral and central processes connect sensory cells of the Corti organ to the central nervous system. The resulting network forms a point-to-point auditory conduction. As a cardiac hormone, brain natriuretic
Lin Ding et al.
Frontiers in immunology, 14, 1139391-1139391 (2023-06-19)
MDSCs express SCHLAFEN 4 (SLFN4) in Helicobacter-infected stomachs coincident with spasmolytic polypeptide-expressing metaplasia (SPEM), a precursor of gastric cancer. We aimed to characterize SLFN4+ cell identity and the role of Slfn4 in these cells. Single-cell RNA sequencing was performed on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service