Skip to Content
MilliporeSigma

371963-M

H-89, Dihydrochloride

A cell-permeable, selective, reversible, ATP-competitive and potent inhibitor of protein kinase A (Ki = 48 nM).

Synonym(s):

H-89, Dihydrochloride, N-[2-(( p-Bromocinnamyl)amino)ethyl]-5-isoquinolinesulfonamide, 2HCl, PKA Inhibitor III, N-[2-((p-Bromocinnamyl)amino)ethyl]-5-isoquinolinesulfonamide, 2HCl, PKA Inhibitor III

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20BrN3O2S · 2HCl
CAS Number:
Molecular Weight:
519.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥99% (HPLC)
Form:
film (crystalline)
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99% (HPLC)

form

film (crystalline)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clearoff-white

solubility

DMSO: 200 mg/mL, water: 25 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NCCNC\C=C\c3ccc(cc3)Br)c1c2c(cncc2)ccc1

InChI

1S/C20H20BrN3O2S/c21-18-8-6-16(7-9-18)3-2-11-22-13-14-24-27(25,26)20-5-1-4-17-15-23-12-10-19(17)20/h1-10,12,15,22,24H,11,13-14H2/b3-2+

InChI key

ZKZXNDJNWUTGDK-NSCUHMNNSA-N

General description

A cell-permeable, selective, reversible, ATP-competitive and potent inhibitor of protein kinase A (Ki = 48 nM). Inhibits other kinases only at much higher concentrations: CaM kinase II (Ki = 29.7 µM), casein kinase I (Ki = 38.3 µM), myosin light chain kinase (Ki = 28.3 µM), protein kinase C (Ki = 31.7 µM), and ROCK-II (IC50 = 270 nM). May be used to discriminate between the effects of PKA and cAMP-regulated guanine-nucleotide-exchange factors (GEFs) such as GEFI, GEFII, or Epac (exchange protein directly activated by cAMP). Enhances radiation-induced apoptosis in the human cell line BM 13674. A 10 mM (1 mg/193 µl) solution of H-89, 2HCl (Cat. No. 371962) in DMSO is also available.

Biochem/physiol Actions

Primary Target
PKA
Target Ki: 48 nM for protein kinase A

Preparation Note

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for at up to 4 months at 4°C.

Other Notes

Leemhuis, J., et al. 2002. J. Pharmacol. Exp. Ther.300, 1000.
Davies, S.P., et al. 2000. Biochem. J.351, 95.
Kawasaki, H., et al. 1998. Science 282, 2275.
de Rooij, J., et al. 1998. Nature 396, 474.
Findik, D., et al. 1995. J. Cell. Biochem.57, 12.
Geilen, C.C. 1992. FEBS Lett.309, 381.
Chijiwa, T., et al. 1990. J. Biol. Chem.265, 5267.
Combest, W.L., et al. 1988. J. Neurochem.51, 1581.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library