Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML1614

Sigma-Aldrich

Rp-8-Br-cGMPS sodium salt

≥98% (HPLC)

Synonym(s):

Rp-8-Bromo-cGMPS sodium salt, Rp-8-Bromoguanosine- 3′, 5′- cyclic monophosphorothioate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10BrN5O6PS · Na
CAS Number:
Molecular Weight:
462.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

color

white to beige

storage temp.

−20°C

SMILES string

O=C1C2=C(N([C@@H]3O[C@H](CO[P@@](O4)([O-])=S)[C@@H]4[C@H]3O)C(Br)=N2)N=C(N)N1.[Na+]

InChI

1S/C10H11BrN5O6PS.Na/c11-9-13-3-6(14-10(12)15-7(3)18)16(9)8-4(17)5-2(21-8)1-20-23(19,24)22-5;/h2,4-5,8,17H,1H2,(H,19,24)(H3,12,14,15,18);/q;+1/p-1/t2-,4-,5-,8-,23-;/m1./s1

InChI key

CHTSSROWUAICIL-GMKUFTCMSA-M

Biochem/physiol Actions

Rp-8-bromo-cGMPS is a cell permeable cGMP analog that blocks cGMP-dependent protein kinase (cGK, G-kinase, Protein Kinase G, PKG). Rp-8-bromo-cGMPS is resistant to hydrolysis by phosphodiesterases and binds cGK without activating it, resulting in competitive inhibition.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fan Xu et al.
Frontiers in pharmacology, 11, 523-523 (2020-05-12)
Ischemic stroke remains the leading cause of death and adult disability. Cerebral ischemic/reperfusion (I/R) injury is caused by ischemic stroke thereafter aggravates overwhelming neuronal apoptosis and even the death of neurons. Of note, hippocampus is more susceptive to cerebral I/R

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service