Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400010

Sigma-Aldrich

Caspase-1 Inhibitor I

The Caspase-1 Inhibitor I, also referenced under CAS 143313-51-3, controls the biological activity of Caspase-1. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-1 Inhibitor I, IL-1β Converting Enzyme (ICE) Inhibitor I, Ac-YVAD-CHO, L-709049

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N4O8
CAS Number:
Molecular Weight:
492.52
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL
water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

N([C@@H](CC(=O)O)C=O)[C@@H](C)C(=O)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)Cc1ccc(cc1)O)C(C)C

InChI

1S/C23H32N4O8/c1-12(2)20(23(35)27-21(33)13(3)24-16(11-28)10-19(31)32)26-22(34)18(25-14(4)29)9-15-5-7-17(30)8-6-15/h5-8,11-13,16,18,20,24,30H,9-10H2,1-4H3,(H,25,29)(H,26,34)(H,31,32)(H,27,33,35)/t13-,16-,18-,20-/m0/s1

InChI key

KQCBPHUOVBKBPQ-SJVNDZIOSA-N

General description

A potent, specific, and reversible inhibitor of caspase-1 (Ki = 200 pM for human recombinant caspase-1), caspase-4, and caspase-5. Strongly inhibits anti-APO-1 induced apoptosis in L929-APO-1 cells.
Interleukin-1β converting enzyme (ICE) is a cysteine protease that cleaves the IL-1β precursor to mature IL-1β. ICE inhibitor I is a potent, specific, and reversible inhibitor of ICE (Ki = 200 pM for human recombinant ICE). Strongly inhibits anti-APO-1 induced apoptosis in L929-APO-1 cells.

Biochem/physiol Actions

Cell permeable: no
Primary Target
caspase-1, human recombinant
Product does not compete with ATP.
Reversible: yes
Target Ki: 200 pM for human recombinant caspase-1

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Tyr-Val-Ala-Asp-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Garcia-Calvo, M., et al. 1998. J. Biol. Chem. 273, 32608.
Hilbi, H., et al. 1998. J. Biol. Chem. 273, 32895.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Los, M., et al. 1995. Nature375, 81.
Walker, N.P.C., et al. 1994. Cell78, 343.
Wilson, K.P., et al. 1994. Nature370, 270.
Thornberry, N.A., et al. 1992. Nature356, 768.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service