Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

369373

Sigma-Aldrich

Pentaerythritol tetrabenzoate

96%

Synonym(s):

Benzoflex S 552, Pentaerythrityl tetrabenzoate, [3-Benzoyloxy-2,2-bis(benzoyloxymethyl)propyl] benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO2CH2)4C
CAS Number:
Molecular Weight:
552.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

102-104 °C (lit.)

functional group

ester
phenyl

SMILES string

O=C(OCC(COC(=O)c1ccccc1)(COC(=O)c2ccccc2)COC(=O)c3ccccc3)c4ccccc4

InChI

1S/C33H28O8/c34-29(25-13-5-1-6-14-25)38-21-33(22-39-30(35)26-15-7-2-8-16-26,23-40-31(36)27-17-9-3-10-18-27)24-41-32(37)28-19-11-4-12-20-28/h1-20H,21-24H2

InChI key

MINJAOUGXYRTEI-UHFFFAOYSA-N

Related Categories

General description

Pentaerythritol tetrabenzoate is an ester of pentaerythritol with benzoic acid. It has been reported as a metal ligand. It has been tested as stationary phase for the quantitative analysis of naphthalene hydrocarbons by gas-liquid chromatography and molecular spectroscopy.

Application

Plasticizer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of pentaerythritol tetrabenzoate.
Bormann LM, et al.
Acta Crystallographica Section C, Structural Chemistry, 49(11), 1982-1985 (1993)
Quantitative analysis of C 10-C 12 naphthalene hydrocarbons using gas chromatography and molecular spectroscopy.
Vaisberg KM, et al.
Chemistry and Technology of Fuels and Oils, 1(9), 736-740 (1965)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service